Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBAS Antibody (OTI1B8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270804H
Description
GBAS Monoclonal specifically detects GBAS in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
GBAS | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NIPSNAP2 | |
Full length recombinant protein of human GBAS (NP_001474) produced in E.coli. | |
0.1 mL | |
2631.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI1B8 | |
Western Blot, Immunohistochemistry | |
glioblastoma amplified sequence, Glioblastoma-amplified sequence, NIPSNAP2NipSnap2,4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2, protein NipSnap homolog 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction