Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBAS Antibody (OTI1B8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245730
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
GBAS Monoclonal specifically detects GBAS in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
GBAS | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:200-1:2000, Immunohistochemistry 1:150, Immunofluorescence 1:50-1:100 | |
glioblastoma amplified sequence, Glioblastoma-amplified sequence, NIPSNAP2NipSnap2,4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2, protein NipSnap homolog 2 | |
Mouse | |
33.6 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
OTI1B8 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
NIPSNAP2 | |
Full length human recombinant protein of human GBAS(NP_001474) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
2631 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction