Learn More
Invitrogen™ GBAS Monoclonal Antibody (OTI1B8)

Description
GBAS Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | GBAS |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1B8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NIPSNAP2 |
| Gene Accession No. | O55126, O75323 |
| Gene Alias | 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2; AV006093; GBAS; glioblastoma amplified sequence; glioblastoma-amplified sequence; nipsnap homolog 2; NIPSNAP2; Protein NipSnap homolog 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.