Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBAS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584582 Supplier Invitrogen™ Supplier No. PA584582
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSLLPRLRTW TSSSNRSRED SWLKSLFVRK VD Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 75%.

GBAS encodes a protein belonging to the NipSnap family, which is believed to participate in vesicular transport. This protein is primarily found in mitochondria and is involved in oxidative phosphorylation. A non-functional version of this gene is located on the long arm of chromosome 2. Multiple isoforms of this gene have been identified due to alternative splicing. NIPSNAP2 is linked to several diseases such as Phosphoserine Phosphatase Deficiency and Mast Syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBAS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NIPSNAP2
Gene Accession No. O75323
Gene Alias 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2; AV006093; GBAS; glioblastoma amplified sequence; glioblastoma-amplified sequence; nipsnap homolog 2; NIPSNAP2; Protein NipSnap homolog 2
Gene Symbols NIPSNAP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NIPSNAP2. Recombinant protein control fragment (Product #RP-103491).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2631
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.