Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GBAS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520655
Description
A suggested positive control is human skeletal muscle tissue lysate. PA5-20655 can be used with blocking peptide PEP-0775.
GBAS encodes a protein belonging to the NipSnap family, which is believed to participate in vesicular transport. This protein is primarily found in mitochondria and is involved in oxidative phosphorylation. A non-functional version of this gene is located on the long arm of chromosome 2. Multiple isoforms of this gene have been identified due to alternative splicing. NIPSNAP2 is linked to several diseases such as Phosphoserine Phosphatase Deficiency and Mast Syndrome.
Specifications
| GBAS | |
| Polyclonal | |
| Unconjugated | |
| NIPSNAP2 | |
| 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2; AV006093; GBAS; glioblastoma amplified sequence; glioblastoma-amplified sequence; nipsnap homolog 2; NIPSNAP2; Protein NipSnap homolog 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14467, 2631, 498174 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O55126, O75323 | |
| NIPSNAP2 | |
| A 14 amino acid peptide near the amino terminus of human NIPSNAP2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction