Learn More
Invitrogen™ GBAS Polyclonal Antibody

Description
A suggested positive control is human skeletal muscle tissue lysate. PA5-20656 can be used with blocking peptide PEP-0776.
Specifications
Specifications
| Antigen | GBAS |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | NIPSNAP2 |
| Gene Accession No. | O55126, O75323 |
| Gene Alias | 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like 2; AV006093; GBAS; glioblastoma amplified sequence; glioblastoma-amplified sequence; nipsnap homolog 2; NIPSNAP2; Protein NipSnap homolog 2 |
| Gene Symbols | NIPSNAP2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.