Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBE1 Antibody (OTI1D11), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270810G
Description
GBE1 Monoclonal specifically detects GBE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
GBE1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GBE1 | |
Full length recombinant protein of human GBE1 (NP_000149) produced in HEK293T cells. | |
0.1 mL | |
2632.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1D11 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
1,4-alpha-glucan-branching enzyme, amylo-(1,4 to 1,6) transglucosidase, amylo-(1,4 to 1,6) transglycosylase, Brancher enzyme, EC 2.4.1.18, GBE, glucan (1,4-alpha), branching enzyme 1, glycogen branching enzyme, Glycogen-branching enzyme | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction