Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBE1 Monoclonal Antibody (OTI3B4)
GREENER_CHOICE

Catalog No. PIMA525023
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525023 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525023 Supplier Invitrogen™ Supplier No. MA525023
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GBE1 Monoclonal Antibody for Western Blot, IHC (P), IP

GBE1 is a glycogen branching enzyme that catalyzes the transfer of alpha-1,4-linked glucosyl units from the outer end of a glycogen chain to an alpha-1,6 position on the same or a neighboring glycogen chain. Branching of the chains is essential to increase the solubility of the glycogen molecule and, consequently, in reducing the osmotic pressure within cells. Highest level of this enzyme are found in liver and muscle. Mutations in this gene are associated with glycogen storage disease IV (also known as Andersen's disease).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBE1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone OTI3B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GBE1
Gene Accession No. Q04446
Gene Alias 1,4-alpha-glucan-branching enzyme; 2310045H19Rik; 2810426P10Rik; amylo-(1,4 to 1,6) transglucosidase; amylo-(1,4 to 1,6) transglycosylase; APBD; Brancher enzyme; D16Ertd536e; GBE; GBE1; glucan (1,4-alpha-), branching enzyme 1; glycogen branching enzyme; glycogen-branching enzyme; GSD4
Gene Symbols GBE1
Host Species Mouse
Immunogen Full length human recombinant protein of GBE1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2632
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.