Learn More
Invitrogen™ GBE1 Polyclonal Antibody

Description
GBE1 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | GBE1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | GBE1 |
| Gene Accession No. | Q04446, Q9D6Y9 |
| Gene Alias | 1,4-alpha-glucan-branching enzyme; 2310045H19Rik; 2810426P10Rik; amylo-(1,4 to 1,6) transglucosidase; amylo-(1,4 to 1,6) transglycosylase; APBD; Brancher enzyme; D16Ertd536e; GBE; GBE1; glucan (1,4-alpha-), branching enzyme 1; glycogen branching enzyme; glycogen-branching enzyme; GSD4 |
| Gene Symbols | GBE1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.