Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117060 Supplier Invitrogen™ Supplier No. PA5117060
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HepG2, NCI-H460, Mouse heart, Mouse liver, Mouse kidney, Rat liver, Rat heart Immunogen sequence: MAAPMTPAAR PEDYEAALNA ALADVPELAR LLEIDPYLKP YAVDFQRRYK QFSQILKNIG ENEGGIDKFS RGYESFGVHR CADGGLYCKE WAPGAEGVFL TGDFNGWNPF SYPYKKLDYG KWELYIPPKQ NKSVLVPHGS KLKVVITSKS GEILYRISPW AKYVVREGDN VNYDWIHWDP EHSYEFKHSR PKKPRSLRIY ESHVGISSHE GKVASYKHFT CNVLPRIKGL GYNCIQLMAI MEHAYYASFG YQITSFFAAS SRYGTPEELQ ELVDTAHSMG IIVLLDVVHS HASKNSADGL.

GBE1 is a glycogen branching enzyme that catalyzes the transfer of alpha-1,4-linked glucosyl units from the outer end of a glycogen chain to an alpha-1,6 position on the same or a neighboring glycogen chain. Branching of the chains is essential to increase the solubility of the glycogen molecule and, consequently, in reducing the osmotic pressure within cells. Highest level of this enzyme are found in liver and muscle. Mutations in this gene are associated with glycogen storage disease IV (also known as Andersen's disease).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBE1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GBE1
Gene Accession No. Q04446, Q9D6Y9
Gene Alias 1,4-alpha-glucan-branching enzyme; 2310045H19Rik; 2810426P10Rik; amylo-(1,4 to 1,6) transglucosidase; amylo-(1,4 to 1,6) transglycosylase; APBD; Brancher enzyme; D16Ertd536e; GBE; GBE1; glucan (1,4-alpha-), branching enzyme 1; glycogen branching enzyme; glycogen-branching enzyme; GSD4
Gene Symbols GBE1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GBE1 (NP_0001493).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2632, 288333, 74185
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.