Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100539 Supplier Invitrogen™ Supplier No. PA5100539
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GBF1.

This gene encodes a member of the Sec7 domain family. The encoded protein is a guanine nucleotide exchange factor that regulates the recruitment of proteins to membranes by mediating GDP to GTP exchange. The encoded protein is localized to the Golgi apparatus and plays a role in vesicular trafficking by activating ADP ribosylation factor 1. The encoded protein has also been identified as an important host factor for viral replication. Multiple transcript variants have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBF1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GBF1
Gene Accession No. Q92538
Gene Alias 1700083E03Rik; AI035702; ARF1GEF; AW049900; BFA-resistant GEF 1; Gbf1; golgi brefeldin A resistant guanine nucleotide exchange factor 1; golgi-specific brefeldin A-resistance factor 1; golgi-specific brefeldin A-resistance guanine nucleotide exchange factor 1; KIAA0248; mKIAA0248
Gene Symbols GBF1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GBF1(Accession Q92538), corresponding to amino acid residues S1773-P1823.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8729
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.