Learn More
Invitrogen™ GBF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120173
Description
Positive test controls include: HeLa. The target is usually found in the following locations: Cytoplasm, Endoplasmic reticulum-Golgi intermediate compartment, Golgi apparatus, Lipid droplet, Membrane, Peripheral membrane protein, cis-Golgi network, trans-Golgi network. Immunogen sequence: MVDKNIYIIQ GEINIVVGAI KRNARWSTHT PLDEERDPLL HSFGHLKEVL NSITELSEIE PNVFLRPFLE VIRSEDTTGP ITGLA.
This gene encodes a member of the Sec7 domain family. The encoded protein is a guanine nucleotide exchange factor that regulates the recruitment of proteins to membranes by mediating GDP to GTP exchange. The encoded protein is localized to the Golgi apparatus and plays a role in vesicular trafficking by activating ADP ribosylation factor 1. The encoded protein has also been identified as an important host factor for viral replication. Multiple transcript variants have been observed for this gene.
Specifications
| GBF1 | |
| Polyclonal | |
| Unconjugated | |
| GBF1 | |
| 1700083E03Rik; AI035702; ARF1GEF; AW049900; BFA-resistant GEF 1; Gbf1; golgi brefeldin A resistant guanine nucleotide exchange factor 1; golgi-specific brefeldin A-resistance factor 1; golgi-specific brefeldin A-resistance guanine nucleotide exchange factor 1; KIAA0248; mKIAA0248 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 107338, 8729 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92538 | |
| GBF1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-85 of human GBF1 (NP_004184.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.