Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBL Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1002124H
Description
GBL Polyclonal antibody specifically detects GBL in Human samples. It is validated for Western BlotSpecifications
GBL | |
Polyclonal | |
Western Blot | |
G protein beta subunit-like, Gable, GbetaL, GBLPop3, Lst8, LST8MGC111011, Mammalian lethal with SEC13 protein 8, mLST8, MTOR associated protein, LST8 homolog (S. cerevisiae), POP3, Protein GbetaL, target of rapamycin complex subunit LST8, TORC subunit LST8, WAT1 | |
The immunogen recognized by this antibody maps to a region between residues 275 and the C-terminus (residue 326) of human G protein Beta subunit-like using the numbering given in entry NP_071767.2 (GeneID 64223). | |
0.1 mL | |
Cancer, Hypoxia, mTOR Pathway | |
64223 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction