Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530796
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530796 Supplier Invitrogen™ Supplier No. PA530796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (91%), Xenopus laevis (88%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

G-protein beta subunit like (GBL protein) is a novel member of WD-40 protein family. It is a highly conserved cytosolic protein consisting of seven WD-40 repeating motifs. LST8 is a functional component of mTOR signaling complex (TORC1) and positively up-regulates the mTOR pathway. It interacts with the kinase domain of mTOR and stabilizes its interaction with raptor. LST8 participates in regulating cell growth and also in nutrient and growth factor-mediated mTOR signaling to S6K1. Reports suggest up-regulated expression of LST8 due to insulin indicating a possible role of LST8 in intracellular events related to insulin-receptor activation including insulin-stimulated glucose transport and glycogen synthesis. Northern Blot analysis detected a ubiquitous expression of LST8 with highest expression in brain and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBL
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene MLST8
Gene Accession No. Q9BVC4
Gene Alias 0610033N12Rik; AA409454; AI505104; AI851821; G beta-like protein; G protein beta subunit-like; Gable; GbetaL; GBL; GBL protein; Lst8; Mammalian lethal with SEC13 protein 8; mLST8; mLST8 / mLST8; MTOR associated protein, LST8 homolog; MTOR associated protein, LST8 homolog (S. cerevisiae); POP3; protein GbetaL; Target of rapamycin complex subunit LST8; TORC subunit gable; TORC subunit LST8; TORC subunit WAT1; transducin (beta)-like 4; WAT1
Gene Symbols MLST8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human mLST8/GBL. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64223
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.