Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588005 Supplier Invitrogen™ Supplier No. PA588005
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNTSPGTVGS DPVILATAGY DHTVRFWQAH SGICTRTVQH QDSQVNALEV TPDRSMIAAA GYQHIRMYDL NSNNPNPIIS YDGVNKNIAS VGFHEDGRWM YTGGEDCTAR IWDLRSRNLQ CQRIFQVNAP INCVCLHPNQ AELIVGDQSG AIHIWDLKTD HNEQLIPEPE VSITSAHIDP DASYMAAVNS TGNCYVWNLT GGIGDEVTQL IPKTKIPAHT RYALQCRFSP DSTLLATCSA DQTCKIWRTS NFSLMTELSI KSGNPGESSR GWMWGCAFSG DSQYIVTASS DNLARLWCVE TGEIKREYGG HQKAVVCLAF NDSVLG; Positive Samples: 293T, HeLa, K-562, 22Rv1, Mouse brain, Mouse kidney, Mouse thymus, Rat brain, Rat testis; Cellular Location: Cytoplasm.

G-protein beta subunit like (GBL protein) is a novel member of WD-40 protein family. It is a highly conserved cytosolic protein consisting of seven WD-40 repeating motifs. LST8 is a functional component of mTOR signaling complex (TORC1) and positively up-regulates the mTOR pathway. It interacts with the kinase domain of mTOR and stabilizes its interaction with raptor. LST8 participates in regulating cell growth and also in nutrient and growth factor-mediated mTOR signaling to S6K1. Reports suggest up-regulated expression of LST8 due to insulin indicating a possible role of LST8 in intracellular events related to insulin-receptor activation including insulin-stimulated glucose transport and glycogen synthesis. Northern Blot analysis detected a ubiquitous expression of LST8 with highest expression in brain and testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MLST8
Gene Accession No. Q9BVC4, Q9DCJ1, Q9Z2K5
Gene Alias 0610033N12Rik; AA409454; AI505104; AI851821; G beta-like protein; G protein beta subunit-like; Gable; GbetaL; GBL; GBL protein; Lst8; Mammalian lethal with SEC13 protein 8; mLST8; mLST8 / mLST8; MTOR associated protein, LST8 homolog; MTOR associated protein, LST8 homolog (S. cerevisiae); POP3; protein GbetaL; Target of rapamycin complex subunit LST8; TORC subunit gable; TORC subunit LST8; TORC subunit WAT1; transducin (beta)-like 4; WAT1
Gene Symbols MLST8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-326 of human MLST8 (NP_0011861031).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56716, 64223, 64226
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.