Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBL Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GBL |
---|---|
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GBL Polyclonal antibody specifically detects GBL in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
GBL | |
Western Blot, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
G protein beta subunit-like, Gable, GbetaL, GBLPop3, Lst8, LST8MGC111011, Mammalian lethal with SEC13 protein 8, mLST8, MTOR associated protein, LST8 homolog (S. cerevisiae), POP3, Protein GbetaL, target of rapamycin complex subunit LST8, TORC subunit LST8, WAT1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-326 of human GBL (NP_001186103.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Polyclonal | |
Rabbit | |
Cancer, Hypoxia, mTOR Pathway | |
PBS with 50% glycerol, pH7.3. | |
64223 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title