Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBP1 Antibody (SR2121), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322422100UL
Description
GBP1 Monoclonal antibody specifically detects GBP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
GBP1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR2121 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
GBP-1, GTP-binding protein 1, Guanine nucleotide-binding protein 1, guanylate binding protein 1, interferon-inducible, guanylate binding protein 1, interferon-inducible, 67kDa, huGBP-1, interferon-induced guanylate-binding protein 1 | |
A synthesized peptide derived from human GBP1 (Uniprot #: P32455) | |
100 μg | |
Immunology, Innate Immunity, Signal Transduction | |
2633 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction