Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBP2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300230AF350
Description
GBP2 Polyclonal specifically detects GBP2 in Human samples. It is validated for ELISA.Specifications
GBP2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
2634 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
Alexa Fluor 350 | |
GBP-2, GTP-binding protein 2, Guanine nucleotide-binding protein 2, guanylate binding protein 2, interferon-inducible, huGBP-2, interferon-induced guanylate-binding protein 2 | |
Produced in rabbits immunized with purified, recombinant GBP2 (AAH73163.1; Met1-Cys588). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction