Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBP2 Antibody (OTI5C8), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270812MFV610
Description
GBP2 Monoclonal antibody specifically detects GBP2 in Human samples. It is validated for Flow CytometrySpecifications
GBP2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
2634 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
OTI5C8 | |
Flow Cytometry | |
GBP-2, GTP-binding protein 2, Guanine nucleotide-binding protein 2, guanylate binding protein 2, interferon-inducible, huGBP-2, interferon-induced guanylate-binding protein 2 | |
Full length human recombinant protein of human GBP2 (NP_004111) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction