Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GBP2 Antibody (OTI5C8), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270812PE
Description
GBP2 Monoclonal specifically detects GBP2 in Human samples. It is validated for Flow Cytometry.Specifications
GBP2 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
GBP2 | |
Full length recombinant protein of human GBP2 (NP_004111) produced in HEK293T cells. | |
0.1 mL | |
2634.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI5C8 | |
Flow Cytometry | |
GBP-2, GTP-binding protein 2, Guanine nucleotide-binding protein 2, guanylate binding protein 2, interferon-inducible, huGBP-2, interferon-induced guanylate-binding protein 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction