Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GBP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112426
Description
GBP2 Polyclonal Antibody for ICC/IF, IHC (P), ELISA
Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.
Specifications
| GBP2 | |
| Polyclonal | |
| Unconjugated | |
| GBP2 | |
| GBP2; GBP-2; GTP-binding protein 2; guanine nucleotide-binding protein 2; guanylate binding protein 2; guanylate binding protein 2, interferon-inducible; guanylate nucleotide binding protein 2; Guanylate-binding protein 1; Guanylate-binding protein 2; interferon-induced guanylate-binding protein 2; isoprenylated 67 kDa protein; mGBP2; mGBP-2; p67 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 2634 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P32456 | |
| GBP2 | |
| Recombinant Human Guanylate-binding protein 2 protein (100-259AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction