Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560078 Supplier Invitrogen™ Supplier No. PA560078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERLLKEGFEN ESKRLQKDIW DIQMRSKSLE PICNIL Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 39%.

Binds GTP, GDP and GMP. Hydrolyzes GTP very efficiently; GDP rather than GMP is the major reaction product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GBP2
Gene Accession No. P32456
Gene Alias GBP2; GBP-2; GTP-binding protein 2; guanine nucleotide-binding protein 2; guanylate binding protein 2; guanylate binding protein 2, interferon-inducible; guanylate nucleotide binding protein 2; Guanylate-binding protein 1; Guanylate-binding protein 2; interferon-induced guanylate-binding protein 2; isoprenylated 67 kDa protein; mGBP2; mGBP-2; p67
Gene Symbols GBP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GBP2. Recombinant protein control fragment (Product #RP-99812).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2634
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.