Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561062
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561062 Supplier Invitrogen™ Supplier No. PA561062
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLEEQEKTLT SKLQEQARVL KERCQGESTQ LQNEIQKLQK TLKKKTKRYM SHK Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 43%.

This gene encodes a member of the guanylate-binding protein (GBP) family. GBPs specifically bind guanine nucleotides (GMP, GDP, and GTP) and contain two of the three consensus motifs found in typical GTP-binding proteins. The encoded protein interacts with a member of the germinal center kinase family. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBP3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GBP3
Gene Accession No. Q9H0R5
Gene Alias GBP3; GBP-3; GTP-binding protein 3; guanine nucleotide-binding protein 3; guanylate binding protein 3; guanylate-binding protein 3; guanylate-binding protein 3 delta C; RP4-531M19.1
Gene Symbols GBP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GBP3. Recombinant protein control fragment (Product #RP-100449).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2635
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.