Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBP4 Polyclonal Antibody, Invitrogen™

Catalog No. FEPA5145314
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145314 20 μL
FEPA5145313 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEPA5145314 Supplier Invitrogen™ Supplier No. PA5145314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Guanylate-binding proteins, such as GBP4, are induced by interferon and hydrolyze GTP to both GDP and GMP.[supplied by OMIM, Dec 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBP4
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene GBP4
Gene Accession No. A4UUI3
Gene Alias AW228052; Gbp3; GBP4; GBP-4; GTP-binding protein 4; guanine nucleotide-binding protein 4; guanylate binding protein 4; guanylate nucleotide binding protein 3; guanylate nucleotide binding protein 4; guanylate-binding protein 4; guanylate-binding protein 4 homolog; macrophage activation 2; Mag-2; mKIAA4245; Mpa2; Mpa-2
Gene Symbols GBP4
Host Species Rabbit
Immunogen Recombinant Mouse GBP4 protein, Met1-Cys304 (Accession #Q61107).
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229900, 310917
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.