Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBP5 Monoclonal Antibody (OTI8B12)
GREENER_CHOICE

Catalog No. PIMA525388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525388 Supplier Invitrogen™ Supplier No. MA525388
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GBP5 Monoclonal Antibody for Western Blot, Flow

The guanylate-binding proteins (GBPs) are a family of interferon-induced GTP-binding proteins function in innate immunity against microbial and viral pathogens. In humans, there are seven GBPs from hGBP1 to hGBP7. Human GBP1 and GBP2 have been shown to exhibit antiviral activity as well as being able to regulate the inhibition of proliferation and invasion of endothelial cells in response to interferon (IFN). GBP5 promotes selective NALP3 inflammasome responses to pathogenic bacteria and soluble but not crystalline inflammasome priming agents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBP5
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI8B12
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GBP5
Gene Accession No. Q96PP8
Gene Alias 5330409J06Rik; GBP5; GBP-5; Gbp5a; GBP-TA antigen; GTP-binding protein 5; Guanine nucleotide-binding protein 5; guanylate binding protein 5; guanylate nucleotide binding protein 5; guanylate-binding protein 5; muGBP-5; UNQ2427/PRO4987
Gene Symbols GBP5
Host Species Mouse
Immunogen Full length human recombinant protein of GBP5 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115362
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.