Learn More
GBP5 Mouse anti-Human, Clone: OTI4D9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502243

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The guanylate-binding proteins (GBPs) are a family of interferon-induced GTP-binding proteins function in innate immunity against microbial and viral pathogens. In humans, there are seven GBPs from hGBP1 to hGBP7. Human GBP1 and GBP2 have been shown to exhibit antiviral activity as well as being able to regulate the inhibition of proliferation and invasion of endothelial cells in response to interferon (IFN). GBP5 promotes selective NALP3 inflammasome responses to pathogenic bacteria and soluble but not crystalline inflammasome priming agents.Specifications
GBP5 | |
Monoclonal | |
Unconjugated | |
GBP5 | |
GBP-5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
115362 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4D9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96PP8 | |
GBP5 | |
Full length human recombit protein of human GBP5 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.