Learn More
Invitrogen™ GBP5 Polyclonal Antibody

Description
Immunogen sequence: TDQALTETEK KKKEAQVKAE AEKAEAQRLA AIQRQNEQMM QERERLHQEQ VRQMEIAKQN WLAEQQKMQE QQMQEQAAQL STTFQAQNRS LLSELQHAQR TVNNDDPCVL L Highest antigen sequence indentity to the following orthologs: Mouse - 41%, Rat - 44%.
Specifications
Specifications
| Antigen | GBP5 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GBP5 |
| Gene Accession No. | Q96PP8 |
| Gene Alias | 5330409J06Rik; GBP5; GBP-5; Gbp5a; GBP-TA antigen; GTP-binding protein 5; Guanine nucleotide-binding protein 5; guanylate binding protein 5; guanylate nucleotide binding protein 5; guanylate-binding protein 5; muGBP-5; UNQ2427/PRO4987 |
| Gene Symbols | GBP5 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.