Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572757
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572757 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572757 Supplier Invitrogen™ Supplier No. PA572757
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GBP6 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Binds GTP, GDP and GMP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBP6
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene GBP6
Gene Accession No. Q6ZN66
Gene Alias AI595338; Gbp6; GBP-6; GTP-binding protein 6; guanine nucleotide-binding protein 6; guanylate binding protein 6; guanylate binding protein family member 6; guanylate binding protein family, member 6; guanylate-binding protein 6; macrophage activation 2 like; Mpa2l
Gene Symbols GBP6
Host Species Rabbit
Immunogen GBP6 antibody for a 15 amino acid synthetic peptide near the carboxy terminus of human GBP6. The immunogen is within the last 50 amino acids of GBP6.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100702, 163351
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.