Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562355 Supplier Invitrogen™ Supplier No. PA562355
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FQENIAQLKK KMERERENYM RELRKMLSHK MKVLEELLTE GFKEIFESLN EEINRLKEQI EAA Highest antigen sequence identity to the following orthologs: Mouse - 54%, Rat - 54%.

Guanylate-binding proteins, such as GBP7, are induced by interferon and hydrolyze GTP to both GDP and GMP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBP7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GBP7
Gene Accession No. Q8N8V2
Gene Alias 9830147J24Rik; GBP4L; Gbp7; GBP-7; GTP-binding protein 7; guanine nucleotide-binding protein 7; guanylate binding protein 7; Guanylate-binding protein 4-like; Guanylate-binding protein 7
Gene Symbols GBP7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GBP7. Recombinant protein control fragment (Product #RP-100778).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388646
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.