Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ GBS Detect™
SDP

Catalog No. p-200005335 Shop All Hardy Diagnostics Products
Click to view available options
No. per Pack:
10
100
This item is not returnable. View return policy
This item is not returnable. View return policy

GBS Detect™ is recommended for the isolation and differentiation by enhanced hemolysis of detection of gamma-hemolytic Group B Streptococcus from GBS enrichment broth procedures.

  • Optically clear polystyrene for distortion free viewing
  • Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding.
  • Packaged in sleeves of 10 plates in a breathable bag that prevents buildup of condensation and excess moisture

Specifications

Color Red
Content And Storage 2°C to 8°C
Format Plate
Product Type Agar
Sterility Sterile
Fill Volume 17 mL
Material Polystyrene
No. per Pack 10
Dimensions (Dia. x H) 100 x 15 mm
Diameter (Metric) 100 mm
Species Streptococcus species
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.