Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBX2 Monoclonal Antibody (385CT10.1.1)
GREENER_CHOICE

Catalog No. PIMA537547
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537547 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537547 Supplier Invitrogen™ Supplier No. MA537547
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GBX2 Monoclonal Antibody for Western Blot

Altered expression of GBX2, part of the homeobox-containing human family of DNA-binding transcription factors, is associated with therapy failure and death in patients with multiple types of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBX2
Applications Western Blot
Classification Monoclonal
Clone 385CT10.1.1
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GBX2
Gene Accession No. P52951
Gene Alias D130058E05Rik; gastrulation and brain-specific homeobox protein 2; gastrulation brain homeobox 2; Gbx2; Gbx-2; Homeobox protein GBX-2; LOW QUALITY PROTEIN: homeobox protein GBX-2; MMoxA; Stimulated by retinoic acid gene 7 protein; Stra7
Gene Symbols GBX2
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 103-131 amino acids from the Central region of human GBX2.
Purification Method Euglobulin Precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2637
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.