Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GBX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518561
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518561 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518561 Supplier Invitrogen™ Supplier No. PA518561
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

Altered expression of GBX2, part of the homeobox-containing human family of DNA-binding transcription factors, is associated with therapy failure and death in patients with multiple types of cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GBX2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene GBX2
Gene Accession No. P52951
Gene Alias D130058E05Rik; gastrulation and brain-specific homeobox protein 2; gastrulation brain homeobox 2; Gbx2; Gbx-2; Homeobox protein GBX-2; LOW QUALITY PROTEIN: homeobox protein GBX-2; MMoxA; Stimulated by retinoic acid gene 7 protein; Stra7
Gene Symbols GBX2
Host Species Goat
Immunogen Synthetic peptide sequence (QGKDESKVEDDPK) corresponding to the internal amino acids of GBX2 (aa 181-194).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2637
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.