Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GBX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566953
Description
Immunogen sequence: AVRGQGKDESK VEDDPKGKEE SFSLESDVDY SSDDNLTGQA AHKEEDPGHA LEET Highest antigen sequence identity to the following orthologs - mouse 96%, rat 96%.
Altered expression of GBX2, part of the homeobox-containing human family of DNA-binding transcription factors, is associated with therapy failure and death in patients with multiple types of cancer.
Specifications
GBX2 | |
Polyclonal | |
Unconjugated | |
GBX2 | |
D130058E05Rik; gastrulation and brain-specific homeobox protein 2; gastrulation brain homeobox 2; Gbx2; Gbx-2; Homeobox protein GBX-2; LOW QUALITY PROTEIN: homeobox protein GBX-2; MMoxA; Stimulated by retinoic acid gene 7 protein; Stra7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2637 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P52951 | |
GBX2 | |
Recombinant Human GBX2. Recombinant protein control fragment (Product #RP-105438). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction