Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gbx2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11849R

Description
Altered expression of GBX2, part of the homeobox-containing human family of DNA-binding transcription factors, is associated with therapy failure and death in patients with multiple types of cancer.Specifications
Gbx2 | |
Polyclonal | |
Unconjugated | |
GBX2 | |
BOS_3885; D130058E05Rik; Gastrulation and brain-specific homeobox protein 2; gastrulation brain homeobox 2; gastrulation brain homeobox 2-like; GBX2; Gbx-2; Homeobox protein GBX-2; LOC100353889; LOW QUALITY PROTEIN: homeobox protein GBX-2; Mmoxa; Stimulated by retinoic acid gene 7 protein; Stra7 | |
Rabbit | |
Protein A | |
RUO | |
2637 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P52951 | |
GBX2 | |
KLH conjugated synthetic peptide derived from human Gbx2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction