Learn More
SGE™ GC Inlet Liner Accessories
SDP
Catalog No. SG0726217 Shop All SGE Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Varian 1078, 1079 Injectors
For Use With (Application):
Graphite Sealing Ring
Catalog No. For Use With (Equipment) For Use With (Application)
SG0726217 Varian 1078, 1079 Injectors Graphite Sealing Ring
1 options
Catalog No. SG0726217 Supplier SGE™ Supplier No. 0726217
Accessories for use with GC inlet liners
  • Viton™ O-Ring—Can be used up to temperatures of 300°C (572°F)
  • Graphite Sealing Rings—Can be used up to temperatures of 450°C (842°F)
Specifications
For Use With (Equipment) Varian 1078, 1079 Injectors
For Use With (Application) Graphite Sealing Ring
Type SGE GC Inlet Liner Accessory