Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GC Monoclonal Antibody (D4), Invitrogen™
Mouse Monoclonal Antibody
$195.80 - $437.00
Specifications
Antigen | GC |
---|---|
Clone | D4 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEMA551072
|
Invitrogen™
MA551072 |
20 μL |
Each for $195.80
|
|
|||||
FEMA551071
|
Invitrogen™
MA551071 |
200 μL |
Each for $437.00
|
|
|||||
Description
GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.Specifications
GC | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
DBP; DBP/ VDBG; DBP/GC; DBP02; DBP-maf; epididymis secretory protein Li 51; Gc; Gc globulin; Gc protein-derived macrophage activating factor; GC, vitamin D binding protein; gc-globulin; GcMAF; Gc-MAF; GRD3; group specific component; Group specific component vitamin D binding protein; Group-specific component; group-specific component (vitamin D binding protein); hDBP; HEL-S-51; V VDB; VDB; VDBG; Vdbp; Vitamin D binding alpha globulin; vitamin D-binding alpha-globulin; vitamin D-binding protein; Vitamin D-binding protein-macrophage activating factor | |
GC | |
IgG1 κ | |
Protein A/G | |
GC |
D4 | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P02774 | |
2638 | |
Recombinant Human DBP protein, Leu209-Asp401 (Accession #P02774). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title