Learn More
Restek™ GC-MS Cleaning Kit
SDP
Catalog No. 06611095 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Description:
With Dremel Tool
Without Dremel Tool
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
06-611-095 With Dremel Tool
06-611-096 Without Dremel Tool
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-611-095 Supplier Restek™ Supplier No. 27194
Add to Cart
Add to Cart
Signs that GC-MS instruments may need cleaning include poor sensitivity, loss of sensitivity at high masses, and high multiplier gain during an auto tune. This kit includes all of the necessary components for cleaning and polishing ion sources.
  • Cleaning Kits are available either with or without battery powered Dremel tool
Specifications
Type GC-MS Cleaning Kit
Description With Dremel Tool
Includes Small Lint-Free Nylon Gloves (2 Pair), Large Lint-Free Nylon Gloves (2 Pair), 9 x 9 Lint-Free Cotton Cloths (10/Pk.), 4 x 6 Micro Mesh Sheets (4/Pk.), Aluminum Oxide (1 kg), Cotton Tip Applicators, Small Tweezers, Large Tweezers, Septum Puller, Battery Operated Dremel Tool, Tool Kit Bag
Quantity 1 Kit