Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GC Recombinant Rabbit Monoclonal Antibody (4P7B3)
GREENER_CHOICE

Catalog No. FEMA551317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551317 Supplier Invitrogen™ Supplier No. MA551317
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: AMDVFVCTYF MPAAQLPELP DVELPTNKDV CDPGNTKVMD KYTFELSRRT HLPEVFLSKV LEPTLKSLGE CCDVEDSTTC FNAKGPLLKK ELSSFIDKGQ ELCADYSENT FTEYKKKLAE RLKAKLPDAT PTELAKLVNK HSDFASNCCS INSPPLYCDS EIDAELKNIL.

GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GC
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4P7B3
Concentration 1.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene GC
Gene Accession No. P02774, P04276, P21614
Gene Alias DBP; DBP/ VDBG; DBP/GC; DBP02; DBP-maf; epididymis secretory protein Li 51; Gc; Gc globulin; Gc protein-derived macrophage activating factor; GC, vitamin D binding protein; gc-globulin; GcMAF; Gc-MAF; GRD3; group specific component; Group specific component vitamin D binding protein; Group-specific component; group-specific component (vitamin D binding protein); hDBP; HEL-S-51; V VDB; VDB; VDBG; Vdbp; Vitamin D binding alpha globulin; vitamin D-binding alpha-globulin; vitamin D-binding protein; Vitamin D-binding protein-macrophage activating factor
Gene Symbols GC
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 305-474 of human Vitamin D Binding protein (NP_000574.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14473, 24384, 2638
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.