Description
- Easy to use
- Forms a reliable seal
- Reusable
- Soft material, forms a soft grip to the capillary column
- Low emissions
Specifications
Specifications
| Material | 100% Graphite |
| Size (Metric) | 1/8 in. |
| Type | GC Union and Nut Replacement Ferrule 1/8 in. |
| Quantity | 10/Pk. |