Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCAP1 Monoclonal Antibody (G2)

Catalog No. MA1724
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1724 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1724 Supplier Invitrogen™ Supplier No. MA1724

Mouse Monoclonal Antibody

MA1-724 detects GCAP-1 from human, non-human primate and bovine samples. This antibody is specific for GCAP-1 and does not react with other isotypes. MA1-724 has been successfully used in Western blot procedures. By Western blot, this antibody detects a ~23 kDa protein representing GCAP-1 protein on bovine rod outer segment membranes. The MA1-724 immunogen is bacterial expressed full length GCAP-1.

Guanylate cyclase-activating protein is a l Ca(2+)-binding protein that upregulates synthesis of cGMP in photoreceptors. The known mammalian GCAPs are more than 90% similar, consisting of 201 to 205 amino acids, and containing 3 identically conserved Ca(2+)-binding sites. The GUCA1A gene, also termed GCAP1, is transcribed into a single 1.7-kb mRNA species detectable only in the retina. In a 4-generation British family with typical clinical features of autosomal dominant cone dystrophy a tyr99-to-cys mutation) in the GUCA1A gene has been identified. Another family with a pro50-to-leu mutation in GUCA1A demonstrated phenotypic variability ranging from mild photophobia to rod-cone dystrophy. The mutant protein could activate guanylate cyclase 1 (GUCY2D) and displayed similar calcium sensitivity to wildtype protein. However, there was a marked increase in the susceptibility to protease degradation and a reduction in the thermal stability of the pro50-to-leu mutation, which may depress cellular concentration and thereby contribute to retinal cell mortality.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCAP1
Applications Western Blot
Classification Monoclonal
Clone G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene Guca1a
Gene Accession No. P43080, P46065
Gene Alias C6orf131; COD3; cone dystrophy 3; CORD14; GC-A; GCAP; GCAP 1; GCAP1; Guanylate cyclase activator 1A; guanylate cyclase activator 1a (retina); guanylate cyclase-activating protein, photoreceptor 1; guanylin 1, retina; guanylyl cyclase-activating protein; guanylyl cyclase-activating protein 1; GUCA; GUCA1; GUCA1A; mGCAP1
Gene Symbols Guca1a
Host Species Mouse
Immunogen Bacterial expressed full length GCAP-1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282243, 2978
Target Species Human, Monkey, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.