Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCAP1 Mouse anti-Rat, Human, Bovine, Porcine, Mouse, Unconjugated, Clone: 6B12, MilliporeSigma™

Catalog No. MABN239625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2396-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN2396-25 Supplier MilliporeSigma Supplier No. MABN239625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GCAP1, clone 6B12, Cat. No. MABN2396, is a highly specific mouse monoclonal antibody that targets Guanylyl cyclase-activating protein 1 and has been tested in Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCAP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6B12
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Guanylyl cyclase-activating protein 1;Guanylate cyclase activator 1A
Gene Symbols Guca1a;Gcap;Gcap1;Guca1
Host Species Mouse
Immunogen GST-fusion protein containing 45 amino acids from the C-terminal region of murine GCAP1. It does not display any homology with GCAP2.
Molecular Weight of Antigen 23 kDa observed; 22.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 6B12 detects Guanylyl cyclase-activating protein 1 in Bovine, Human, Mouse, Porcine, and Rat. It targets an epitope within 45 amino acids from the C-terminal region.
Target Species Rat, Human, Bovine, Pig, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.