Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582201
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582201 Supplier Invitrogen™ Supplier No. PA582201
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EYVAALSLVL KGKVEQKLRW YFKLYDVDGN GCIDRDELLT IIQAIRAINP CSDTTMTAEE FTDTVFSKID VNGDGELSLE EFIEGVQKDQ MLLDTLTRSL DLTRIVRRLQ NGEQDEEGAD EAAEAAG Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 88%.

Guanylate cyclase-activating protein is a l Ca(2+)-binding protein that upregulates synthesis of cGMP in photoreceptors. The known mammalian GCAPs are more than 90% similar, consisting of 201 to 205 amino acids, and containing 3 identically conserved Ca(2+)-binding sites. The GUCA1A gene, also termed GCAP1, is transcribed into a single 1.7-kb mRNA species detectable only in the retina. In a 4-generation British family with typical clinical features of autosomal dominant cone dystrophy a tyr99-to-cys mutation) in the GUCA1A gene has been identified. Another family with a pro50-to-leu mutation in GUCA1A demonstrated phenotypic variability ranging from mild photophobia to rod-cone dystrophy. The mutant protein could activate guanylate cyclase 1 (GUCY2D) and displayed similar calcium sensitivity to wildtype protein. However, there was a marked increase in the susceptibility to protease degradation and a reduction in the thermal stability of the pro50-to-leu mutation, which may depress cellular concentration and thereby contribute to retinal cell mortality.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCAP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Guca1a
Gene Accession No. P43080
Gene Alias C6orf131; COD3; cone dystrophy 3; CORD14; GC-A; GCAP; GCAP 1; GCAP1; Guanylate cyclase activator 1A; guanylate cyclase activator 1a (retina); guanylate cyclase-activating protein, photoreceptor 1; guanylin 1, retina; guanylyl cyclase-activating protein; guanylyl cyclase-activating protein 1; GUCA; GUCA1; GUCA1A; mGCAP1
Gene Symbols Guca1a
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GUCA1A. Recombinant protein control fragment (Product #RP-105700).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2978
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.