Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCAP2 Antibody (A1), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1205422AF405
Description
GCAP2 Monoclonal specifically detects GCAP2 in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GCAP2 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GUCA1B | |
Bacterial expressed full length GCAP-2. | |
0.1 mL | |
Signal Transduction, Vision | |
2979 | |
Bovine | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
A1 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
DKFZp686E1183, GCAP 2, GCAP2guanylate cyclase-activating protein, photoreceptor 2, Guanylate cyclase activator 1B, guanylate cyclase activator 1B (retina), guanylyl cyclase-activating protein 2, GUCA2, RP48 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
This is specific for GCAP 2 and does not react with other isotypes. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction