Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545026 Supplier Thermo Scientific Supplier No. PA545026
Add to Cart
Add to Cart

GCAP2 Polyclonal Antibody

GCAP2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen GCAP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GUCA1B
Gene Accession No. Q9UMX6
Gene Alias BC018258; DKFZp686E1183; GCAP 2; GCAP2; GCAP-2; GCAP2 (retina); guanylate cyclase activator 1B; guanylate cyclase activator 1B (retina); guanylate cyclase-activating protein, photoreceptor 2; Guanylin 2; guanylyl cyclase-activating protein 2; GUCA 2; GUCA1B; GUCA2; Retinal guanylyl cyclase activator protein p24; RP48
Gene Symbols GUCA1B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GUCA1B (aa 34-83).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2979
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.