Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584255 Supplier Invitrogen™ Supplier No. PA584255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GQEFSWEEAE AAGEIDVAEL QEWYKKFVME CPSGTLFMHE FKRFFKVTDD EEASQYVEGM FRAFDKNG Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 90%.

Guanylate cyclase-activating proteins (GCAPs) are calcium binding proteins that belong to the calmodulin superfamily. GCAPs without calcium are responsible for activation of photoreceptor guanylate cyclase during light adaptation. Studies have shown that the addition or subtraction of Ca2+ results in major conformational changes and the activation/deactivation of GCAP-1 and GCAP-2. It has been demonstrated that both GCAP-1 and 2 act on guanylate cyclase similarly and have approximately 50% homology between the two forms. GCAP-1 is predominantly localized in the photoreceptor outer segments, while GCAP-2 is found in retina samples.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCAP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GUCA1B
Gene Accession No. Q9UMX6
Gene Alias BC018258; DKFZp686E1183; GCAP 2; GCAP2; GCAP-2; GCAP2 (retina); guanylate cyclase activator 1B; guanylate cyclase activator 1B (retina); guanylate cyclase-activating protein, photoreceptor 2; Guanylin 2; guanylyl cyclase-activating protein 2; GUCA 2; GUCA1B; GUCA2; Retinal guanylyl cyclase activator protein p24; RP48
Gene Symbols GUCA1B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GUCA1B. Recombinant protein control fragment (Product #RP-108413).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.