Learn More
Invitrogen™ GCAT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590717
Description
Immunogen sequence: KAHKYRYRHL DMADLEAKLQ EAQKHRLRLV ATDGAFSMDG DIAPLQEICC LASRYGALVF MDECHATGFL GPTGRGTDEL LGVMDQVTII NSTLGKALGG ASGGYTTGPG PLVSLLRQRA RPYLFSNSLP PAVVGCASKA LDLLMGSNTI VQSMAAKTQR FRSKMEAAGF TISGASHPIC PVMLGDARLA SRMADDMLKR GIFVIGFSYP VVPKGKARIR VQISAVHSEE DIDRCVEAFV EVGRLHGALP; Positive Samples: Mouse eye; Cellular Location: Mitochondrion, Nucleus.
The degradation of L-threonine to glycine consists of a two-step biochemical pathway involving the enzymes L-threonine dehydrogenase and 2-amino-3-ketobutyrate coenzyme A ligase. L-Threonine is first converted into 2-amino-3-ketobutyrate by L-threonine dehydrogenase. GCAT is the second enzyme in this pathway, which then catalyzes the reaction between 2-amino-3-ketobutyrate and coenzyme A to form glycine and acetyl-CoA. The enzyme is considered a class II pyridoxal-phosphate-dependent aminotransferase.
Specifications
GCAT | |
Polyclonal | |
Unconjugated | |
Gcat | |
2-amino-3-ketobutyrate coenzyme A ligase; 2-amino-3-ketobutyrate coenzyme A ligase, mitochondrial; 2-amino-3-ketobutyrate-CoA ligase; AI526977; AKB ligase; aminoacetone synthase; aminoacetone synthetase; aminotransferase; glycine acetyltransferase; aminoacetone synthetase; KBL; GCAT; Glycine acetyltransferase; glycine C-acetyltransferase; glycine C-acetyltransferase (2-amino-3-ketobutyrate coenzyme A ligase); glycine C-acetyltransferase (2-amino-3-ketobutyrate-coenzyme A ligase); KBL; Unknown (protein for MGC:139551) | |
Rabbit | |
Affinity chromatography | |
RUO | |
23464, 26912, 366959 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.67 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O75600, O88986 | |
Gcat | |
Recombinant fusion protein containing a sequence corresponding to amino acids 170-419 of human GCAT (NP_0551061). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.