Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCAT Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2828940.1ML
Description
GCAT Polyclonal specifically detects GCAT in Human samples. It is validated for Western Blot.Specifications
GCAT | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23464 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
2-amino-3-ketobutyrate-CoA ligase, AKB ligase, Aminoacetone synthase, EC 2.3.1, EC 2.3.1.29, Glycine acetyltransferase, glycine C-acetyltransferase, glycine C-acetyltransferase (2-amino-3-ketobutyrate coenzyme A ligase), KBL2-amino-3-ketobutyrate coenzyme A ligase, mitochondrial, MGC23053 | |
The immunogen is a synthetic peptide directed towards the middle region of human GCAT | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction