Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554381 Supplier Invitrogen™ Supplier No. PA554381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GDPADTSSSD SLTQALQLAA ANEPTFFLYA EQLARKEVEI TSLRKQKHRL EVEVHQLQDR LLEEGERHRE EVAALQSHIE KNIRDQSREG ANLEYLKNII YRFLTLPDSL GRQQTLTAIL TILHFSPEEK QVIMRLPT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

The protein encoded by this gene is a peripheral membrane protein. It is sensitive to brefeldin A. This encoded protein contains a GRIP domain which is thought to be used in targeting. It may play a role in the organization of trans-Golgi network subcompartment involved with membrane transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GCC1
Gene Accession No. Q96CN9, Q9D4H2
Gene Alias 4932417P04Rik; Gcc1; GCC1P; GCC88; golgi coiled coil 1; Golgi coiled coil protein 1; golgi coiled-coil 1; golgi coiled-coil protein 1; GRIP and coiled-coil domain containing 1; GRIP and coiled-coil domain-containing protein 1; peripheral membrane Golgi protein
Gene Symbols GCC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GCC1. Recombinant protein control fragment (Product #RP-92812).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100361513, 74375, 79571
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.