Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GCC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598724
Description
GCC1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene is a peripheral membrane protein. It is sensitive to brefeldin A. This encoded protein contains a GRIP domain which is thought to be used in targeting. It may play a role in the organization of trans-Golgi network subcompartment involved with membrane transport.
Specifications
GCC1 | |
Polyclonal | |
Unconjugated | |
GCC1 | |
4932417P04Rik; Gcc1; GCC1P; GCC88; golgi coiled coil 1; Golgi coiled coil protein 1; golgi coiled-coil 1; golgi coiled-coil protein 1; GRIP and coiled-coil domain containing 1; GRIP and coiled-coil domain-containing protein 1; peripheral membrane Golgi protein | |
Rabbit | |
Protein G | |
RUO | |
79571 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96CN9 | |
GCC1 | |
Recombinant Human GRIP and coiled-coil domain-containing protein 1 protein (4-775AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction