Learn More
Epredia™ Lab Vision™ GCDFP-15 (Gross Cystic Disease Fluid Protein-15) Ab-1, Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: 23A3
Isotype: IgG2a/κ
Species Reactivity: Human and Rat. Others not known.
Immunogen: Recombinant protein encoding the excreted domain of human GCDFP-15
Molecular Weight: 15 kDa
Positive Control: Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | GCDFP-15 (Gross Cystic Disease Fluid Protein-15) Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 23A3 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein encoding the excreted domain of human GCDFP15 |
| Quantity | 12 mL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.